Order Entry
Italy
ContactUsLinkComponent
Tetramethylammonium bromide, Supelco®
Toxic
Tetramethylammonium bromide, Supelco®
Catalog # 87708-10G
Supplier:  Merck
CAS Number:  
undefined
Toxic
Tetramethylammonium bromide, Supelco®
Catalog # 87708-10G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    10 g
  • Seq:
    3236

Specifications

About this item

 :
 :154.05 g/mol
Boiling Pt:∼360 °C (1013 hPa)
Density:1.56 g/cm³ (20 °C)
ADR:6.1,II
 :M-Clarity™ quality level = MQ100
EINECS:200-581-2
UN:2811
 :GHS06
CAS Number:64-20-0
Formula:C₄H₁₂BrN
MDL Number:MFCD00011626