
(1-Propyl)triphenylphosphonium bromide ≥99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:6228-47-3
:MFCD00011843
Formula:C₂₁H₂₂BrP
:385.28 g/mol
Melting Pt:236…240 °C
EINECS:228-330-2
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together