
(1-Hexyl)triphenylphosphonium bromide for synthesis, Sigma-Aldrich®
Catalog # 8.42007.0025
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Solubility:174 g/l (20 °C)
- Melting Point:198 - 203 °C
- Molar Mass:427.37 g/mol
Specification Test Results
About this item
:
:427.36 g/mol
Melting Pt:198…200 °C
:M-Clarity™ quality level = MQ200
EINECS:225-301-6
:GHS07
Storage Temperature:Ambient
CAS Number:4762-26-9
Formula:H₃C(CH₂)₅P(C₆H₅)₃Br
MDL Number:MFCD00011860