
(1-Hexadecyl)triphenylphosphonium bromide ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:14866-43-4
:
Formula:H₃C(CH₂)₁₅P(C₆H₅)₃Br
MDL Number:MFCD00051858
:567.63 g/mol
Melting Pt:99…101 °C
EINECS:238-939-5
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together