
(1-Butyl)triphenylphosphonium bromide 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1779-51-7
:
Formula:H₃C(CH₂)₃P(C₆H₅)₃Br
MDL Number:MFCD00011855
:399.31 g/mol
Melting Pt:240…243 °C
Storage Temperature:Ambient
EINECS:217-219-4
UN:3464
ADR:6.1,III
:GHS06
Specification Test Results
- Appearance:White to off-white Powder
- Infrared spectrum:Conforms
- Melting point:238 to 243 °C
- Titration Argentometric:≥98.5 %
- Specific optical rotation:-9.5° to -12.5° (20 °C, 589 nm) (c=1, methanol)
Specification Test Results
Frequently Bought Together