

Dimethyldichlorosilane for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:75-78-5
:
Formula:C₂H₆Cl₂Si
MDL Number:MFCD00000491
:129.06 g/mol
Boiling Pt:70 °C (1013 hPa)
Melting Pt:–76 °C
Density:1.0745 g/cm³ (20 °C)
Flash Pt:–16 °C
Storage Temperature:Ambient
UN:1162
ADR:3 (8),II
:M-Clarity™ quality level = MQ200
EINECS:200-901-0
REACH:01-2119437250-51
:GHS02
:GHS07
Specification Test Results
- Solubility:(20 °C) (rigorous decomposition)
- Melting Point:-76 °C
- Molar Mass:129.06 g/mol
- Boiling Point:70 °C (1013 hPa)
- Vapor Pressure:- 200 hPa (20 °C)
- Flash Point:-5 °C
- Refractive Index:1.4038 (20 °C, 589 nm)
- Explosion Limit:1.75 - 48.5 %(V)
- Density:1.07 g/cm3 (25 °C)
- Ignition Point:425 °C
Specification Test Results
Related Information
Frequently Bought Together