
(3-Aminopropyl)trimethoxysilane per sintesi, Sigma-Aldrich®
Catalog # 8.18550.0250
Supplier: Merck
CAS Number:
Specifications
- Tipo di conf.:bottiglia in vetro
- Conf:250 ml
- Seq:2590
Specifications
Specification Test Results
- Solubility:(20 °C) (decomposition)
- Molar Mass:179.29 g/mol
- Boiling Point:194 °C (1013 hPa)
- Flash Point:90 °C
- Density:1.02 g/cm3 (20 °C)
- pH:9 (20 g/l, H2O, 20 °C)
- Ignition Point:295 °C
Specification Test Results
About this item
:
Boiling Pt:194 °C (1013 hPa)
Density:1,0265 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
:179.29 g/mol
EINECS:237-511-5
:GHS07
Storage Temperature:Ambiente
CAS Number:13822-56-5
Formula:H₂NCH₂CH₂CH₂Si(OCH₃)₃
MDL Number:MFCD00008206