Silicon(II) oxide ≥99.9% (metals basis), sputtering target, Ø 50.8 mm (2.0 in), Thickness: 6.35 mm (0.250 in)
Catalog # 41093.KS
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:1
- Seq:0001
Specifications
About this item
CAS Number:10097-28-6
:
Formula:SiO
MDL Number:MFCD00151536
Merck Index:12,08640
Boiling Pt:1880 °C
Melting Pt:>1702 °C
Density:2.13
:44.08 g/mol
Storage Temperature:Ambient
EINECS:233-232-8