Order Entry
Italy
ContactUsLinkComponent
 
Aluminium oxide, Durmax® for metallography, 30 minutes
Catalog # 20981.265
Supplier:  ALUMINES DURMAX
CAS Number:  
Aluminium oxide, Durmax® for metallography, 30 minutes
Catalog # 20981.265
Supplier:  ALUMINES DURMAX
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    500 mL
  • Seq:
    2725

Specifications

About this item

 :
 :101.96 g/mol
Merck Index:13,00355
Boiling Pt:2980 °C (1013 hPa)
Melting Pt:2045 °C
Density:3.94 g/cm³ (20 °C)
EINECS:215-691-6
REACH:01-2119529248-35
CAS Number:1344-28-1
 :11092-32-3
Formula:Al₂O₃
MDL Number:MFCD00003424