Order Entry
Italy
ContactUsLinkComponent
Boron nitride ≥99.99% (metals basis), sputtering target, Ø 76.2 mm (3.0 in), Thickness: 3.18 mm (0.125 in)
Boron nitride ≥99.99% (metals basis), sputtering target, Ø 76.2 mm (3.0 in), Thickness: 3.18 mm (0.125 in)
  41054.KS
 :  
Boron nitride ≥99.99% (metals basis), sputtering target, Ø 76.2 mm (3.0 in), Thickness: 3.18 mm (0.125 in)
  41054.KS
 :  
  • Pk:
    1
  • Seq:
    0001
 :10043-11-5
 :
 :BN
 :MFCD00011317
 :24.82 g/mol
 :13,01336
 :2527 °C
 :2.15 g/cm³ (20 °C)
 :Ambient
 :233-136-6