Barium sulphate for white standard DIN 5033 (Blanc fixe), Supelco®
Catalog # 1.01748.0250
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:250 g
- Seq:3545
Specifications
About this item
:
:233.39 g/mol
Merck Index:13,00997
Boiling Pt:1600 °C (1013 hPa)
Melting Pt:1345 °C
Density:4.5 g/cm³ (25 °C)
:M-Clarity™ quality level = MQ100
Storage Temperature:Ambient
EINECS:231-784-4
CAS Number:7727-43-7
Formula:BaSO₄
MDL Number:MFCD00003455