Barium sulphate 99%, precipitated
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7727-43-7
:
Formula:BaSO₄
MDL Number:MFCD00003455
:233.39 g/mol
Merck Index:14,994
Boiling Pt:1600 °C (1013 hPa)
Melting Pt:1580 °C (dec.)
Density:4.5
Storage Temperature:Ambient
EINECS:231-784-4
Related Information
Frequently Bought Together