Order Entry
Italy
ContactUsLinkComponent
 
Magnesium oxide ≥99.95% (metals basis excluding Ca), sputtering target, Ø 50.8 mm (2.0 in), Thickness: 3.18 mm (0.125 in)
Catalog # 41072.KS
CAS Number:  
Magnesium oxide ≥99.95% (metals basis excluding Ca), sputtering target, Ø 50.8 mm (2.0 in), Thickness: 3.18 mm (0.125 in)
Catalog # 41072.KS
CAS Number:  

Specifications

  • Pk:
    1
  • Seq:
    0001

Specifications

About this item

CAS Number:1309-48-4
 :
Formula:MgO
MDL Number:MFCD00011109
 :40.3 g/mol
Merck Index:13,05700
Boiling Pt:3600 °C (1013 hPa)
Melting Pt:2800 °C
Density:3.58 g/cm³ (25 °C)
Storage Temperature:Ambient
EINECS:215-171-9