
Calcium hydride ∼92%, coarse powder
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7789-78-8
:
Formula:H₂Ca
MDL Number:MFCD00010897
:42.09 g/mol
Merck Index:14,1672
Melting Pt:600 °C (dec.)
Density:1.900
Storage Temperature:Ambient
EINECS:232-189-2
UN:1404
ADR:4.3,I
:GHS02
Related Information
Frequently Bought Together