Order Entry
Italy
Orders LinkContactUsLinkComponent
 

Barium chromate ≥98%

Health
Oxidizing
Irritant
Nome del fornitore:   Thermo Fisher Scientific
Barium chromate ≥98%
Nome del fornitore:  Thermo Fisher Scientific
Certificates
Barium chromate ≥98%
Barium chromate ≥98%

2 Options Available Below

Product Details & Documents

Purity: 98% CAS No.: 10294-40-3 Molecular Formula: BaCrO4 Formula Weight: 253.33 Physical Form: Powder Density: 4.50 Appearance: Yellow Odour: Odourless Solubility: Insoluble in water, acetic acid, chromic acid Merck Reference: 14,972 UN No.: 1479 MDL No.: MFCD00014180

  • CAS Number:10294-40-3
  • Formula:BaCrO₄
  • MDL Number:MFCD00014180
  • Molecular Weight:253.32 g/mol
  • Merck Index:14,972
  • Melting Pt:1400 °C
  • Density:4.50
  • Storage Temperature:Ambient
  • EINECS:233-660-5
  • UN:1479
  • ADR:5.1,III
  •  :Density: 4,5 g/cm³ (25 °C)
Product Family Options

Product Information