Order Entry
Italy
ContactUsLinkComponent
Barium carbonate ≥99.0%, AnalaR NORMAPUR® analytical reagent
Barium carbonate ≥99.0%, AnalaR NORMAPUR® analytical reagent
Supplier:  VWR Chemicals
Certificates
undefined
Barium carbonate ≥99.0%, AnalaR NORMAPUR® analytical reagent
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Assay:
    Min. 99.0 %
  • Alkali hydroxides+carbonates(as Ba(OH)2):
    Max. 0.015 %
  • Heavy metals (as Pb):
    Max. 10 ppm
  • Insolubility in hydrochloric acid:
    Max. 0.015 %
  • Oxidising substances (as NO3):
    Max. 50 ppm
  • S (Sulphur):
    Max. 10 ppm
  • Ca (Calcium):
    Max. 0.02 %
  • Cl (Chloride):
    Max. 20 ppm
  • Fe (Iron):
    Max. 10 ppm
  • K (Potassium):
    Max. 50 ppm
  • Na (Sodium):
    Max. 0.02 %
  • Sr (Strontium):
    Max. 0.3 %

Specification Test Results

Frequently Bought Together