
Barium carbonate ≥99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:513-77-9
:
Formula:BaCO₃
MDL Number:MFCD00003448
:197.34 g/mol
Merck Index:14,969
Boiling Pt: Dec.
Melting Pt:811 °C
Density:4.43
Storage Temperature:Ambient
EINECS:208-167-3
UN:1564
ADR:6.1,III
:GHS07
Related Information
Frequently Bought Together