
Chromium(III) potassium sulphate dodecahydrate, black crystals ACS
Catalog # ICNA0219403190
Supplier: MP Biomedicals
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:500 g
- Seq:3645
Specifications
Specification Test Results
- Assay:98.0 - 102.0%
- Insoluble Matter:<0.01%
- Chloride (Cl):<0.002%
- Aluminum (Al):<0.02%
- Iron (Fe):<0.01%
- Ammonium (NH4):<0.01%
Specification Test Results
About this item
:
:499.4 g/mol
Merck Index:13,02250
Melting Pt:89 °C (lit.)
Density:1.89 g/cm³ ((20 °C))
EINECS:233-401-6
:GHS07
Storage Temperature:Ambient
CAS Number:7788-99-0
Formula:CrK(SO₄)₂·12H₂O
MDL Number:MFCD00149917