
Rubidium sulphate ≥99% (metals basis)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7488-54-2
:
Formula:O₄Rb₂S
MDL Number:MFCD00011190
:267 g/mol
Boiling Pt:1708 °C (1013 hPa)
Melting Pt:1050 °C
Density:3.613
Storage Temperature:Ambient
EINECS:231-301-7
:GHS07
Related Information
Frequently Bought Together