


Potassium hexachloroplatinate(IV) (∼40% Pt)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:16921-30-5
:
Formula:K₂PtCl₆
MDL Number:MFCD00011389
Merck Index:13,07720
Melting Pt:250 °C (dec.)
Density:3,344 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:240-979-3
UN:2923
ADR:8,III
:485.99 g/mol
:GHS06
:GHS08
:GHS05
Specification Test Results
- Appearance:Yellow to light orange Powder and/or chunks
- Platinum (Pt):39.4 to 40.6 % (Pt)
Specification Test Results
Frequently Bought Together