Order Entry
Italy
Orders LinkContactUsLinkComponent
 

Potassium hydroxide

1
Catalog # CAYM760713-1
Corrosive
Irritant
Supplier:   Cayman Chemical
Potassium hydroxide
Catalog # CAYM760713-1
Supplier:  Cayman Chemical
CAS Number:  1310-58-3
Potassium hydroxide

1

CAS Number:  1310-58-3
Price
21,60 €
Log in for availability
1 * 1 pezzi
Product Details & Documents
Synonyms:  Potash caustic, Caustic potash
  • CAS Number:1310-58-3
  • Formula:KOH
  • MDL Number:MFCD00003553
  • Molecular Weight:56.11 g/mol
  • Merck Index:12,07806
  • Boiling Pt:1320 °C (1013 hPa)
  • Melting Pt:360 °C
  • Density:2.04 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:215-181-3
  • UN:1813
  • ADR:8,II
  • REACH:01-2119487136-33
Specifications

Specifications

  • Pk:
    1

Specifications