Order Entry
Italy
Orders LinkContactUsLinkComponent
Potassium hydroxide ≥85%, soluble

Potassium hydroxide ≥85%, soluble

Corrosive
Irritant
Potassium hydroxide ≥85%, soluble
Certificates
Potassium hydroxide ≥85%, soluble
Potassium hydroxide ≥85%, soluble

3 Options Available Below

Product Details & Documents

Purity: 85% CAS No.: 1310-58-3 Molecular Formula: HKO Formula Weight: 56.11 Odour: Odourless pH: 13,5 Melting Point: 360° Boiling Point: 1320-1324° Merck Reference: 14,7640 UN No.: 1813 MDL No.: MFCD00003553

Synonyms:  Potash caustic, Caustic potash
  • CAS Number:1310-58-3
  • Formula:KOH
  • MDL Number:MFCD00003553
  • Molecular Weight:56.11 g/mol
  • Merck Index:14,7640
  • Boiling Pt:1320…1324 °C
  • Melting Pt:360 °C
  • Density:2.044
  • Storage Temperature:Ambient
  • EINECS:215-181-3
  • UN:1813
  • ADR:8,II
  • REACH:01-2119487136-33
Product Family Options

Product Information