Order Entry
Italy
ContactUsLinkComponent
 
Potassium chloride
Catalog # HACHS21M001
Supplier:  Hach
CAS Number:  
Potassium chloride
Catalog # HACHS21M001
Supplier:  Hach
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    15 g
  • Seq:
    3266

Specifications

About this item

 :
 :74.55 g/mol
Merck Index:14,07621
Boiling Pt:1420 °C (1013 hPa)
Melting Pt:773 °C
Density:1.98 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-211-8
CAS Number:7447-40-7
Formula:KCl
MDL Number:MFCD00011360