Order Entry
Italy
Orders LinkContactUsLinkComponent
Dantron Impurity Standard, Neat

Dantron Impurity Standard, Neat

Dantron Impurity Standard, Neat, Glass bottle, 100 mg
Catalog # PROOBP429
Dantron Impurity Standard, Neat
Catalog # PROOBP429
CAS Number:  
Dantron Impurity Standard, Neat

Dantron Impurity Standard, Neat, Glass bottle, 100 mg

CAS Number:  
Price
260,00 €
Log in for availability
1 * 100 mg
Product Details & Documents
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 mg
  • Description:
    Dantron Impurity Standard, Neat
  • Cat. No.:
    PROOBP429
  • Formula:
    C₁₄H₈O₄
  • Description:
    Dantron
  • Appearance:
    Neat
  • Synonym(s):
    1,8-Dihydroxyanthracene-9,10-dione, Dithranol Imp. B (EP), Danthron, 1,8-Dihydroxy-9,10-anthraquinone, 1,8-Dihydroxyanthraquinone, 1,8-Dioxyanthraquinone, Altan, Antrapurol, Chrysazin, Danthrone, Dantron, Diaquone, Dionone, Dorbane, Istin, Istizin, Laxanorm, Laxanthreen, Laxipur, Laxipurin, Modane, NSC 38626, NSC 646568, NSC 7210, Zwitsalax, Dithranol Impurity B
  • Molecular weight:
    240.21 g/mol
  • CAS No.:
    117-10-2

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.