Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Italy
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Chloro-9,10-anthraquinone
2-Chloro-9,10-anthraquinone
2-Chloro-9,10-anthraquinone
PROOERM-FC022
:
LGC Standards PROMOCHEM
:
2-Chloro-9,10-anthraquinone
PROOERM-FC022
:
LGC Standards PROMOCHEM
:
Pk:
1
Seq:
0001
:
:
242.66 g/mol
:
292 °C (30 mmHg)
:
207…210 °C
:
1 g/cm³ (20 °C)
:
205-010-0
:
GHS07
:
131-09-9
:
C₁₄H₇ClO₂
:
MFCD00001227