1-Methylpiperazine ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:109-01-3
:
Formula:C₅H₁₂N₂
MDL Number:MFCD00005966
:100.16 g/mol
Boiling Pt:137…139 °C
Melting Pt:–6 °C
Density:0.903
Flash Pt:34 °C (93 °F)
Storage Temperature:Ambient
EINECS:203-639-5
UN:2734
ADR:8,II
:GHS06
:GHS05
:GHS02
Related Information
Frequently Bought Together