Order Entry
Italy
ContactUsLinkComponent
2-Thiobarbituric acid, Supelco®
2-Thiobarbituric acid, Supelco®
Catalog # 1.08180.0025
Supplier:  Merck
CAS Number:  
2-Thiobarbituric acid, Supelco®
Catalog # 1.08180.0025
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Solubility:
    (20 °C) slightly soluble
  • Melting Point:
    235 °C
  • Molar Mass:
    144.14 g/mol
  • Bulk Density:
    210 kg/m3
  • pH:
    1.7 - 1.9 (10 g/l, H2O, 20 °C)

Specification Test Results

About this item

 :
 :144.15 g/mol
Boiling Pt:419 °C (1013 hPa)
Melting Pt:235 °C
Density:1.661 g/cm³ (20 °C)
 :M-Clarity™ quality level = MQ100
Storage Temperature:Ambient
EINECS:207-985-8
CAS Number:504-17-6
Formula:C₄H₄N₂O₂S
MDL Number:MFCD00006674