Order Entry
Italy
ContactUsLinkComponent
Cyclohexanecarbonyl chloride for synthesis, Sigma-Aldrich®
Corrosive
Irritant
Cyclohexanecarbonyl chloride for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
Corrosive
Irritant
Cyclohexanecarbonyl chloride for synthesis, Sigma-Aldrich®
Supplier:  Merck

Specification Test Results

  • Solubility:
    (20 °C) (decomposition)
  • Molar Mass:
    146.62 g/mol
  • Boiling Point:
    180 - 181 °C (1013 hPa)
  • Flash Point:
    66 °C
  • Density:
    1.08 g/cm3 (20 °C)

Specification Test Results

Related Information

Frequently Bought Together