

Cyclohexanecarbonyl chloride 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:2719-27-9
:
Formula:C₇H₁₁ClO
MDL Number:MFCD00001456
Boiling Pt:184 °C (1013 hPa)
Melting Pt:29 °C
Density:1.09 g/cm³
Flash Pt:66 °C
Storage Temperature:Ambient
EINECS:220-322-7
ADR:8,II
:146.62 g/mol
UN:3265
:GHS07
:GHS05
Specification Test Results
- Appearance:Clear colorless to straw yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4680 to 1.4700 (20°C, 589 nm)
- GC:≥97.5 %
- Titration RCOCl:≥97.5 %
- Titration RCOOH:≤2 %
- Free acid (HCl):Traces
Specification Test Results
Frequently Bought Together