

p-Benzoquinone
Catalog # EHERC10537000
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:0,25 g
- Seq:3029
Specifications
About this item
:
Merck Index:12,08259
Boiling Pt:∼174 °C (1013 hPa)
Melting Pt:112…114 °C
Density:1.318 g/cm³ (20 °C)
Flash Pt:77 °C
Storage Temperature:Ambient
EINECS:203-405-2
UN:2587
ADR:6.1,II
:108.1 g/mol
:GHS06
:GHS09
CAS Number:106-51-4
Formula:C₆H₄O₂
MDL Number:MFCD00001591