Order Entry
Italy
ContactUsLinkComponent
Benzo[a]pyrene
Health
Environment
Benzo[a]pyrene
Catalog # EHERC20635000
Supplier:  EHRENSTORFER
CAS Number:  
Health
Environment
Benzo[a]pyrene
Catalog # EHERC20635000
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    10 mg
  • Seq:
    2275

Specifications

About this item

 :
Merck Index:12,01134
Boiling Pt:495 °C (1013 hPa)
Melting Pt:176…178 °C
Density:1.351 g/cm³ (20 °C)
EINECS:200-028-5
UN:2811
ADR:6.1,III
 :252.32 g/mol
 :GHS08
 :GHS09
CAS Number:50-32-8
Formula:C₂₀H₁₂
MDL Number:MFCD00003602