Order Entry
Italy
ContactUsLinkComponent
 
2,2'-Dihydroxybiphenyl for synthesis, Sigma-Aldrich®
  8.20154.0050
 :  Merck
 :  
2,2'-Dihydroxybiphenyl for synthesis, Sigma-Aldrich®
  8.20154.0050
 :  Merck
 :  

 

  • Pack type:
    Plastic bottle
  • Pk:
    50 g
  • Seq:
    3368

 

 

  • Solubility:
    (20 °C) insoluble
  • Melting Point:
    106 - 109 °C
  • Molar Mass:
    186.2 g/mol
  • Boiling Point:
    314 - 316 °C
  • Flash Point:
    154 °C
  • Ignition Point:
    324 °C
  • Synonyms:
    o,o'-Biphenol, 2,2'-Dihydroxydiphenyl
  • Hill Formula:
    C12H10O2
  • Chemical Formula:
    2-(HO)C6H4C6H4-2-(OH)

 

 

 :
 :186.21 g/mol
 :315 °C (1013 hPa)
 :106…109 °C
 :1.342 g/cm³ (20 °C)
 :M-Clarity™ quality level = MQ200
 :217-303-0
 :1806-29-7
 :C₁₂H₁₀O₂
 :MFCD00002210