2,2'-Dimethoxybiphenyl ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:4877-93-4
:
Formula:C₁₄H₁₄O₂
MDL Number:MFCD00067073
:214.26 g/mol
Boiling Pt:307.5 °C (1013 hPa)
Melting Pt:152…156 °C
Density:1.268 g/cm³ (25 °C)
Storage Temperature:Ambient
Frequently Bought Together