3-Phenylaniline
Catalog # EHERC10173030
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:0,1 g
- Seq:3020
Specifications
About this item
:
:169.23 g/mol
Boiling Pt:342 °C (1013 hPa)
Boiling Pt:342 °C (1013 hPa)
Melting Pt:33 °C
CAS Number:2243-47-2
Formula:C₁₂H₁₁N
MDL Number:MFCD00047846