Supelco® Organic Reference Standard, PCB No 29
Catalog # 31093-20MG
Supplier: Merck
CAS Number:
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Pack type:Glass bottle for solids
- Pk:20 mg
- Standards:Analytical standard
- CAS No.:15862-07-4
- Description:PCB No 29
- Storage conditions:2...30 °C
- Hazard class description:H373 - H410
- Shipping conditions:STD
- MDL No.:MFCD00055248
- M-Clarity quality level:MQ100
- Synonym(s):2,4,5-Trichlorobiphenyl; 2,4,5-PCB
- Molecular weight:257,54
- Flash point:Not applicable
Specifications
About this item
- GHS signal: GHS08, GHS09
Caution: P273 - P314