2,3',4,4',5,5'-Hexachlorobiphenyl (PCB No. 167)
Catalog # EHERC20016700
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:10 mg
- Seq:2275
Specifications
About this item
:
:360.88 g/mol
Boiling Pt:383 °C (1013 hPa)
Melting Pt:125…127 °C
UN:3432
ADR:9,II
CAS Number:52663-72-6
Formula:C₁₂H₄Cl₆