Order Entry
Italy
ContactUsLinkComponent
 
 
2,5-Di-tert-butylhydroquinone for synthesis, Sigma-Aldrich®
 :  Merck
 
 
2,5-Di-tert-butylhydroquinone for synthesis, Sigma-Aldrich®
 :  Merck

 

  • Solubility:
    (20 °C) insoluble
  • Melting Point:
    214 - 217 °C
  • Molar Mass:
    222.32 g/mol
  • Ignition Point:
    420 °C

 

Frequently Bought Together