Order Entry
Italy
ContactUsLinkComponent
Phenol red, powder, Bioreagent cell culture reagent, Sigma-Aldrich®
Phenol red, powder, Bioreagent cell culture reagent, Sigma-Aldrich®
Certificates
undefined
Phenol red, powder, Bioreagent cell culture reagent, Sigma-Aldrich®
Certificates

Specification Test Results

  • Appearance (Colour):
    Dark Red to Dark Brown
  • Appearance (Form):
    Powder
  • Solubility (Colour)
    100 mg plus 100 ml 1 N NaOH:
    Dark Red
  • Infrared spectrum:
    Conforms to Structure
  • pH
    6.8 (yellow), 8.2 (red):
    Conforms
  • Cell Culture Test:
    Pass
  • Recommended Retest Period:
    3 years

Specification Test Results

Related Information

Frequently Bought Together