Phenol red ACS
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:143-74-8
:
Formula:C₁₉H₁₄O₅S
MDL Number:MFCD00003552
Merck Index:13,07329
Melting Pt:>300 °C
:354.38 g/mol
EINECS:205-609-7
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Red Powder
- Infrared spectrum:Conforms
- Visual transition interval:From pH 6.8 (yellow) to pH 8.2 (red)
- Clarity of solution:Passes test
Specification Test Results
Frequently Bought Together