2,4-Dihydroxybenzophenone for synthesis, Sigma-Aldrich®
Catalog # 8.18652.0250
Supplier: Merck
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:250 g
- Seq:3545
Specifications
Specification Test Results
- Solubility:(20 °C) insoluble
- Melting Point:143 - 146 °C
- Molar Mass:214.22 g/mol
- Bulk Density:750 kg/m3
- Boiling Point:194 °C
- Density:1.32 g/cm3 (20 °C)
- pH:5 (10 g/l, H2O, 20 °C) (slurry)
Specification Test Results
About this item
:
:214.22 g/mol
Merck Index:13,01107
Boiling Pt:391 °C (1013 hPa)
Melting Pt:139…140 °C
Density:0.696 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
EINECS:205-029-4
CAS Number:131-56-6
Formula:C₁₃H₁₀O₃
MDL Number:MFCD00002277