Order Entry
Italy
Orders LinkContactUsLinkComponent
 

Chlorophenol red, pure indicator grade

Irritant
Chlorophenol red, pure indicator grade
Certificates
Chlorophenol red, pure indicator grade

2 Options Available Below

Product Details & Documents
Synonyms:  3,3-Bis(3-chloro-4-hydroxyphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide, 4,4'-(3H-2,1-Benzoxathiol-3-ylidene)bis[2-chlorophenol] S,S-dioxide
  • CAS Number:4430-20-0
  • Formula:C₁₉H₁₂Cl₂O₅S
  • MDL Number:MFCD00005877
  • Molecular Weight:423.27 g/mol
  • Melting Pt:261 °C
  • Storage Temperature:Ambient
  • EINECS:224-619-2

Documents

Specifications for Product Family

Specification Test Results

  • Appearance:
    Green to red-brown to brown Powder
  • Infrared spectrum:
    Conforms
  • Loss on drying:
    ≤8 % (110°C)
  • Solubility:
    (0.1 % in ethanol) Clear brown-yellow to dark orange
  • UV Lambda max:
    572 to 576 nm (in 0.1 N NaOH)
  • Specific absorbance A (1%/1cm):
    ≥1000 (at Lambda max) On dried substance
  • Visual transition interval:
    from pH 4.8 (yellow) to pH 6.4 (purple)
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    10 g
    Log in for availability
    Log in for availability
    1 * 10 g
    1 * 10 g - 66,80 €
    66,80 €
    1 * 10 g
       66,80 €

  • Glass bottle
    50 g
    Log in for availability
    Log in for availability
    1 * 50 g
    1 * 50 g - 223,00 €
    223,00 €
    1 * 50 g
       223,00 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.