Jump to:
Product Details & Documents
Synonyms: 4,4'-(3H-2,1-Benzoxathiol-3-ylidene)bis[2-bromophenol] S,S-dioxide
- CAS Number:2800-80-8
- Formula:C₁₉H₁₂Br₂O₅S
- MDL Number:MFCD00023014
- Molecular Weight:512.18 g/mol
- Melting Pt:230 °C
- :M-Clarity™ quality level = MQ200
- EINECS:220-538-1
Specifications
Specification Test Results
- Solubility:(20 °C) slightly soluble
- Melting Point:230 °C (decomposition)
- Molar Mass:512.18 g/mol
- Bulk Density:770 kg/m3
Specifications
- Pack type:Glass bottle
- Pk:5 g



