Bromophenol red, pure
Supplier: Thermo Fisher Scientific
About this item
Synonyms: 4,4'-(3H-2,1-Benzoxathiol-3-ylidene)bis[2-bromophenol] S,S-dioxide
- CAS Number:2800-80-8
- Formula:C₁₉H₁₂Br₂O₅S
- MDL Number:MFCD00023014
- Molecular Weight:512.18 g/mol
- Melting Pt:230 °C
- EINECS:220-538-1
2 Options Available Below
Product Details & Documents
Synonyms: 4,4'-(3H-2,1-Benzoxathiol-3-ylidene)bis[2-bromophenol] S,S-dioxide
- CAS Number:2800-80-8
- Formula:C₁₉H₁₂Br₂O₅S
- MDL Number:MFCD00023014
- Molecular Weight:512.18 g/mol
- Melting Pt:230 °C
- EINECS:220-538-1
Documents
Specifications for Product Family
Specification Test Results
- Appearance:Red-brown to brown Powder
- Infrared spectrum:Conforms
- Loss on drying:≤5 % (110°C)
- Solubility:(1 % in ethanol) Clear
- Visual transition interval:from pH 5.2 (yellow-red) to pH 7.0 (purple)
Product Family Options
Product Information
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



