

p-Toluenesulphonyl chloride 99+%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:98-59-9
:
Formula:C₇H₇ClO₂S
MDL Number:MFCD00007450
:190.65 g/mol
Merck Index:13,09612
Boiling Pt:134 °C (10 mmHg)
Melting Pt:67…69 °C
Density:1,49 g/cm³ (20 °C)
Flash Pt:128 °C
Storage Temperature:Ambient
EINECS:202-684-8
UN:3261
ADR:8,II
:GHS05
:GHS07
Specification Test Results
- Appearance:White Crystalline powder
- Infrared spectrum:Conforms
- Melting point:67 to 69 °C
- Assay:≥99.0 %
- Impurity:≤0.3 % Total free acid (as toluenesulfonic acid)
Specification Test Results
Frequently Bought Together