Order Entry
Italy
ContactUsLinkComponent
 
 
Benzyl alcohol, EMPROVE® EXPERT Ph. Eur., BP, JP, NF, special quality (max. 0.05% benzaldehyde), SAFC®
 
undefined
 
Benzyl alcohol, EMPROVE® EXPERT Ph. Eur., BP, JP, NF, special quality (max. 0.05% benzaldehyde), SAFC®
 

 

  • Assay (GC):
    98.0 - 100.5 %
  • Identity (IR):
    conforms
  • Appearance of solution (nephelometric):
    conforms
  • Clarity of solution:
    conforms
  • Color of solution:
    conforms
  • Acidity:
    conforms
  • Refractive index (n 20/D):
    1.538 - 1.541
  • Density (d 20 °C/20 °C):
    1.043 - 1.049
  • Peroxide value:
    ≤ 5.0
  • Related substances (GC) (Benzaldehyde (Impurity A)):
    ≤ 0.05 %
  • Related substances (GC) (Cyclohexylmethanol (Impurity B)):
    ≤ 0.10 %
  • Related substances (GC) (Sum of peaks with a relative retention less than that of benzyl alcohol):
    ≤ 0.02 %
  • Related substances (GC) (Sum of peaks with a relative retention greater than that of benzyl alcohol):
    ≤ 0.20 %
  • Related substances (GC) (disregard limit):
    ≤ 0.0001 %
  • Benzene (GC):
    ≤ 0.0002 %
  • Chlorobenzene (GC):
    ≤ 0.01 %
  • Toluene (GC):
    ≤ 0.01 %
  • Other residual solvents (ICH Q3C):
    excluded by manufacturing process
  • Evaporation residue:
    ≤ 0.050 %
  • Water:
    ≤ 0.1 %
  • Bacterial endotoxins:
    ≤ 2.5 I.U./ml
  • Total aerobic microbial count (TAMC):
    ≤ 100 CFU/g
  • Total combined yeasts/moulds count (TYMC):
    ≤ 10 CFU/g
  • Bile-tolerant gram-negative bacteria (absent in 1 g):
    passes test
  • Candida albicans (absent in 1 g):
    passes test
  • Escherichia coli (absent in 1 g):
    passes test
  • Pseudomonas aeruginosa (absent in 1 g):
    passes test
  • Salmonella (absent in 10 g):
    passes test
  • Staphylococcus aureus (absent in 1 g):
    passes test

 

Frequently Bought Together