Order Entry
Italy
ContactUsLinkComponent
 
 
Benzyl alcohol 98.0-100.5% NF, Multi-Compendial, J.T.Baker®
 :  Avantor
 
undefined
 
Benzyl alcohol 98.0-100.5% NF, Multi-Compendial, J.T.Baker®
 :  Avantor
 

 

  • Meets B.P. Chemical Specifications:
  • Meets E.P. Chemical Specifications:
  • Meets N.F. Requirements:
  • Meets J.P. Chemical Specifications:
  • GMP Manufactured Product:
  • CAUTION: For Manufacturing, processing or repackaging:
  • Bulk Pharmaceutical Chemical:
    98.0 - 100.5 %
  • NF - Assay (C₆H₅CH₂OH):
    98.0 - 100.5 %
  • NF - Clarity of Solution:
    Passes Test
  • NF - Color of Solution:
    Passes Test
  • NF - Identification:
    Passes Test
  • NF - Peroxide Value:
    ≤5
  • NF - Benzaldehyde and Other Related Substances - Benzaldehyde:
    ≤0.15 %
  • NF - Related Substances - Cyclohexylmethanol:
    ≤0.10 %
  • of Benzyl Alcohol:
    ≤0.04 %
  • NF - Benzaldehyde and Other Related Substances - Total Other Peaks
    ≤0.04 %
  • NF - Benzaldehyde and Other Related Substances - Total Other Peaks:
    ≤0.3 %
  • Greater Than RT of Benzyl Alcohol:
    ≤0.3 %
  • NF - Refractive Index, n²ºD:
    1.538 - 1.541
  • NF - Acidity:
    Passes Test
  • NF - Residue on Evaporation:
    ≤0.05 %
  • NF - Benzene:
    ≤2 ppm
  • Solubility:
    Passes Test
  • EP/BP - Assay (C₆H₅CH₂OH):
    98.0 - 100.5 %
  • EP/BP - Identification:
    Passes Test
  • EP/BP - Acidity:
    Passes Test
  • EP/BP - Appearance of Solution:
    Passes Test
  • EP/BP - Refractive Index, n²º^D:
    1.538 - 1.541
  • EP/BP - Benzaldehyde and Other Related Substances - Benzaldehyde:
    ≤0.15 %
  • EP/BP - Related Substances - Cyclohexylmethanol:
    ≤0.10 %
  • ≤0.04 %
  • EP/BP - Benzaldehyde and Other Related Substances - Total Other Peaks:
    ≤0.04 %
  • >RT OF BENZYL ALCOHOL:
    ≤0.3 %
  • EP/BP - Peroxide Value:
    ≤5
  • EP/BP - Residue on Evaporation:
    ≤0.05 %
  • JP - Assay (C₆H₅CH₂OH):
    98.0 - 100.5 %
  • JP - Identification:
    Passes Test
  • JP - Refractive Index, n²º^D:
    1.538 - 1.541
  • JP - Specific Gravity at 20°/20°C:
    1.043 - 1.049
  • JP - Clarity and Color of Solution:
    Passes Test
  • JP - Acidity:
    Passes Test
  • JP - Benzaldehyde and Other Related Substances - Benzaldehyde:
    ≤0.15 %
  • JP - Related Substances - Cyclohexylmethanol:
    ≤0.10 %
  • JP - Benzaldehyde and Other Related Substances - Total Other Peaks
    ≤0.04 %
  • JP - Benzaldehyde and Other Related Substances - TOTAL OTHER PEAKS >RT:
    ≤0.3 %
  • JP - Peroxide Value:
    ≤5
  • JP - Residue on Evaporation:
    ≤0.05 %

 

 

 
 
Frequently Bought Together