Order Entry
Italy
ContactUsLinkComponent
α,α,α-Trifluoro-p-cresol for synthesis, Sigma-Aldrich®
Irritant
α,α,α-Trifluoro-p-cresol for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
Irritant
α,α,α-Trifluoro-p-cresol for synthesis, Sigma-Aldrich®
Supplier:  Merck

Specification Test Results

  • Melting Point:
    46 - 48 °C
  • Molar Mass:
    162.11 g/mol
  • Boiling Point:
    73 - 75 °C (11 hPa)
  • Flash Point:
    83 °C

Specification Test Results

Related Information

Frequently Bought Together