

α,α,α-Trifluoro-m-cresol 98+%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:98-17-9
:
Formula:C₇H₅F₃O
MDL Number:MFCD00002299
:162.11 g/mol
Boiling Pt:178 °C (1013 hPa)
Melting Pt:–1.8 °C
Density:1.33 g/cm³
Flash Pt:73 °C
Storage Temperature:Ambient
EINECS:202-645-5
UN:1760
ADR:8,II
:GHS05
:GHS07
Specification Test Results
- Appearance:Clear colorless to yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4570 to 1.4600 (20°C, 589 nm)
- GC:≥98.0 %
Specification Test Results
Frequently Bought Together