
2',5'-Dimethoxyacetophenone 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1201-38-3
:
Formula:C₁₀H₁₂O₃
MDL Number:MFCD00008728
:180.2 g/mol
Boiling Pt:155…158 °C (11 mmHg)
Melting Pt:18…20 °C
Density:1.13 g/cm³
Flash Pt:>112 °C
EINECS:214-858-0
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Clear yellow to yellow-green Liquid
- Infrared spectrum:Conforms
- Refractive index:1.5421 to 1.5441 (20°C, 589 nm)
- GC:≥98.5 %
Specification Test Results
Frequently Bought Together