


6-Chloro-o-cresol ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:87-64-9
:
Formula:C₇H₇ClO
MDL Number:MFCD00020000
:142.58 g/mol
Boiling Pt:75…76 °C (12 mmHg)
Melting Pt:0 °C
Density:1.201
Flash Pt:72 °C (161 °F)
Storage Temperature:Ambient
EINECS:201-760-8
UN:2669
ADR:6.1,II
:GHS06
:GHS05
:GHS07
Related Information
Frequently Bought Together